Learn More
Biomatik Corporation Rat Interleukin 12, IL-12/P40 ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKC40308
Rat Interleukin 12, Il-12/P40 Elisa Kit for detecting Interleukin 12, Il-12/P40 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.