Learn More
Biomatik Corporation Mouse Vascular Endothelial cell Growth Factor B, VEGF-B ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40310
Encompass_Preferred
Mouse Vascular Endothelial Cell Growth Factor B, Vegf-B Elisa Kit for detecting Vascular Endothelial Growth Factor B in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 0.195pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49766; Vegfb. Target Synonyms: Vegfb; VrfVascular endothelial growth factor B; VEGF-B; VEGF-related factor; VRF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9145