Learn More
Biomatik Corporation Mouse Noradrenaline, NA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40338
Encompass_Preferred
Mouse Noradrenaline, Na Elisa Kit for detecting Noradrenaline, Na in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.5-300pg/ml. Sensitivity: 7.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9157