Learn More
Biomatik Corporation Rat Prostaglandin E2, PG-E2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40358
Encompass_Preferred
Rat Prostaglandin E2, Pg-E2 Elisa Kit for detecting Prostaglandin E2, Pg-E2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.5-100pg/ml. Sensitivity: 0.25pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9163