Learn More
Biomatik Corporation Human Tissue Inhibitor of Matrix Metalloprotease-1 (TIMP-1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40364
Encompass_Preferred
Human Tissue Inhibitor Of Matrix Metalloprotease-1 (Timp-1) Elisa Kit for detecting Timp Metallopeptidase Inhibitor 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.39-25ng/ml. Sensitivity: 0.434ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01033; TIMP1. Target Synonyms: Clgi; Collagenase inhibitor; Collagenase inhibitor; Human; EPA; EPO; Erythroid Potentiating Activity; Erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Human Collagenase Inhibitor; Metalloproteinase inhibitor 1; Metalloproteinase inhibitor 1 precursor; OTTHUMP00000023214; TIMP 1; TIMP; TIMP metallopeptidase inhibitor 1; TIMP-1; Timp1; TIMP1 protein; TIMP1_HUMAN; Tissue Inhibitor of Metalloproteinase 1; Tissue inhibitor of metalloproteinases 1; Tissue inhibitor of metalloproteinases. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9167