Learn More
Biomatik Corporation Human High mobility group protein B1, HMGB-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40405
Encompass_Preferred
The Human High mobility group protein B1, HMGB-1 ELISA Kit allows for quantitative determination of high-mobility group box 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 78 pg/mL-5000 pg/mL. Sensitivity: 19.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09429. Restriction: For Research Use Only.
Catalog No. 50-149-9191