Learn More
Biomatik Corporation Mouse High mobility group protein B1, HMGB-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40407
Encompass_Preferred
Mouse High Mobility Group Protein B1, Hmgb-1 Elisa Kit for detecting High-Mobility Group Box 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P63158; Hmgb1. Target Synonyms: Hmgb1; Hmg-1; Hmg1High mobility group protein B1; High mobility group protein 1; HMG-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9193