Learn More
Biomatik Corporation Mouse transferrin receptor, TFR ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40435
Encompass_Preferred
Mouse Transferrin Receptor, Tfr Elisa Kit for detecting Transferrin Receptor (P90, Cd71) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q62351; Tfrc. Target Synonyms: Tfrc; Trfr; Transferrin receptor protein 1; TR; TfR; TfR1; Trfr; CD antigen CD71. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9208