Learn More
Biomatik Corporation Human cardiotrophln 1, CT-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40444
Encompass_Preferred
The Human cardiotrophln 1, CT-1 ELISA Kit allows for quantitative determination of cardiotrophin 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q16619. Restriction: For Research Use Only.
Catalog No. 50-149-9213