Learn More
Biomatik Corporation Fish Tri-iodothyronine, T3 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40453
Encompass_Preferred
The Fish Tri-iodothyronine, T3 ELISA Kit allows for quantitative determination of Tri-iodothyronine, T3 in serum, plasma and tissue homogenates. Species Reactivity: Fish. Detection Range: 0.5 ng/mL-8 ng/mL. Sensitivity: 0.5 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9216