Learn More
Biomatik Corporation Human dipeptidyl peptidase Ⅳ, DPPⅣ ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40462
Encompass_Preferred
The Human dipeptidyl peptidase Ⅳ, DPPⅣ ELISA Kit allows for quantitative determination of dipeptidyl-peptidase 4 in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P27487. Restriction: For Research Use Only.
Catalog No. 50-149-9221