Learn More
Biomatik Corporation Chicken Interleukin 6, IL-6 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40468
Encompass_Preferred
The Chicken Interleukin 6, IL-6 ELISA Kit allows for quantitative determination interleukin 6 (interferon, beta 2) in serum, plasma, tissue homogenates. Species Reactivity: Chicken. Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q90YI0. Restriction: For Research Use Only.
Catalog No. 50-149-9223