Learn More
Biomatik Corporation Chicken Interferon γ, IFN-γ ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40469
Encompass_Preferred
The Chicken Interferon γ, IFN-γ ELISA Kit allows for quantitative determination interferon, gamma in serum, plasma, tissue homogenates. Species Reactivity: Chicken. Detection Range: 3.125 pg/mL-800 pg/mL. Sensitivity: 3.125 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P49708. Restriction: For Research Use Only.
Catalog No. 50-149-9224