Learn More
Biomatik Corporation Human Cholesteryl ester transfer protein, CETP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40475
Encompass_Preferred
The Human Cholesteryl ester transfer protein, CETP ELISA Kit allows for quantitative determination of cholesteryl ester transfer protein, plasma in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.78 ng/mL-50 ng/mL. Sensitivity: 0.195 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P11597. Restriction: For Research Use Only.
Catalog No. 50-149-9228