Learn More
Biomatik Corporation Rat Cardiac Troponin (cTn-) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40482
Encompass_Preferred
Rat Cardiac Troponin I (Ctn-I) Elisa Kit for detecting Cardiac Troponin I(Ctn-I) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P23693; Tnni3. Target Synonyms: Tnni3; Ctni; Tni; Troponin I; cardiac muscle; Cardiac troponin I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9230