Learn More
Biomatik Corporation Human glial fibrillary acidic protein, GFAP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40483
Encompass_Preferred
The Human glial fibrillary acidic protein, GFAP ELISA Kit allows for quantitative determination of glial fibrillary acidic protein in serum, plasma and tissue homogenates. Species Reactivity: Human. Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P14136. Restriction: For Research Use Only.
Catalog No. 50-149-9231