Learn More
Biomatik Corporation Rat Myeloperoxidase (MPO) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKC40506
Rat Myeloperoxidase (Mpo) Elisa Kit for detecting Myeloperoxidase, Mpo in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: D3ZGE2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.