Learn More
Biomatik Corporation Human Catenin beta-1 (CTNNB1/CTNNB/OK/SW-cl.35/PRO2286) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40547
Encompass_Preferred
The Human Catenin beta-1 (CTNNB1/CTNNB/OK/SW-cl.35/PRO2286) ELISA Kit allows for quantitative determination of catenin (cadherin-associated protein), beta 1, 88kDa in serum, plasma, cell culture supernates, tissue homogenates and cell lysates. Species Reactivity: Human. Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P35222. Restriction: For Research Use Only.
Catalog No. 50-149-9255