Learn More
Biomatik Corporation Human Peptide YY ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40556
Encompass_Preferred
The Human Peptide YY ELISA Kit allows for quantitative determination of peptide YY in serum, plasma, urine, tissue homogenates. Species Reactivity: Human. Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P10082. Restriction: For Research Use Only.
Catalog No. 50-149-9257