Learn More
Biomatik Corporation Human Glucagon, GC ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40585
Encompass_Preferred
The Human Glucagon, GC ELISA Kit allows for quantitative determination of glucagon in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 12.9 pg/mL-200 pg/mL. Sensitivity: 4.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01275. Restriction: For Research Use Only.
Catalog No. 50-149-9262