Learn More
Biomatik Corporation Human β-hexosaminidase A, β-Hex A ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40627
Encompass_Preferred
The Human β-hexosaminidase A, β-Hex A ELISA Kit allows for quantitative determination of β-hexosaminidase A, β-Hex A. Species Reactivity: Human. Detection Range: 1.56 mIU/mL-100 mIU/mL. Sensitivity: 0.39 mIU/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9278