Learn More
Biomatik Corporation Human islet amyloid polypeptide, IAPP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40670
Encompass_Preferred
The Human islet amyloid polypeptide, IAPP ELISA Kit allows for quantitative determination of islet amyloid polypeptide in serum, plasma, urine. Species Reactivity: Human. Detection Range: 47 pg/mL-3000 pg/mL. Sensitivity: 11.75 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P10997. Restriction: For Research Use Only.
Catalog No. 50-149-9280