Learn More
Biomatik Corporation Human dopamine-β-hydroxylase, DBH ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40671
Encompass_Preferred
The Human dopamine-β-hydroxylase, DBH ELISA Kit allows for quantitative determination of dopamine beta-hydroxylase (dopamine beta-monooxygenase) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 2.5 ng/mL-500 ng/mL. Sensitivity: 1.25 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09172. Restriction: For Research Use Only.
Catalog No. 50-149-9281