Learn More
Biomatik Corporation Rat omentin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40680
Encompass_Preferred
Rat Omentin Elisa Kit for detecting Omentin in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9284