Learn More
Biomatik Corporation Bovine Interleukin 2, IL-2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40692
Encompass_Preferred
The Bovine Interleukin 2, IL-2 ELISA Kit allows for quantitative determination interleukin 2 in serum, plasma, tissue homogenates. Species Reactivity: Bovine. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P05016. Restriction: For Research Use Only.
Catalog No. 50-149-9289