Learn More
Biomatik Corporation Goat Tumor necrosis factor α, TNF-α ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40693
Encompass_Preferred
The Goat Tumor necrosis factor α, TNF-α ELISA Kit allows for quantitative determination of tumor necrosis factor (TNF superfamily, member 2) in serum, plasma, tissue homogenates. Species Reactivity: Goat. Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P13296. Restriction: For Research Use Only.
Catalog No. 50-149-9290