Learn More
Biomatik Corporation Monkey Interferon , IFN- ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40732
Encompass_Preferred
Monkey Interferon Α, Ifn-Α Elisa Kit for detecting Interferon Α, Ifn-Α in serum, urine, cell culture supernates, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: IFN-alpha,interferon alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9299