Learn More
Biomatik Corporation Chicken Interferon α, IFN-α ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40740
Encompass_Preferred
The Chicken Interferon α, IFN-α ELISA Kit allows for quantitative determination interferon α, IFN-α in serum, plasma, tissue homogenates. Species Reactivity: Chicken. Detection Range: 3.12 pg/mL-200 pg/mL. Sensitivity: 0.78 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9301