Learn More
Biomatik Corporation Human angiotensin Ⅱ receptor 1, ANGⅡR-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40787
Encompass_Preferred
The Human angiotensin Ⅱ receptor 1, ANGⅡR-1 ELISA Kit allows for quantitative determination of angiotensin II receptor, type 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 46.88 pg/mL-3000 pg/mL. Sensitivity: 11.72 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P30556. Restriction: For Research Use Only.
Catalog No. 50-149-9320