Learn More
Biomatik Corporation Rabbit cardiac troponin , cTn-ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40795
Encompass_Preferred
Rabbit Cardiac Troponin I, Ctn-Ielisa Kit for detecting Troponin I Type 3 (Cardiac) in serum, plasma, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 47-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02646; TNNI3. Target Synonyms: TNNI3; Troponin I; cardiac muscle; Cardiac troponin I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9323