Learn More
Biomatik Corporation Human peroxisome proliferative activated receptor gama coactivator 1 alpha, PPARGC1 ELISA Kit, 96 tests
Catalog No. 501499332
Encompass_Preferred
The Human peroxisome proliferative activated receptor gama coactivator 1 alpha, PPARGC1 ELISA Kit allows for quantitative determination of peroxisome proliferator-activated receptor gamma, coactivator 1 alpha in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Human. Detection Range: 125 pg/mL-8000 pg/mL. Sensitivity: 31.25 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9UBK2. Restriction: For Research Use Only.
Catalog No. 50-149-9332 Supplier Biomatik Corporation Supplier No. EKC40809
The Human peroxisome proliferative activated receptor gama coactivator 1 alpha, PPARGC1 ELISA Kit allows for quantitative determination of peroxisome proliferator-activated receptor gamma, coactivator 1 alpha in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Human. Detection Range: 125 pg/mL-8000 pg/mL. Sensitivity: 31.25 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9UBK2. Restriction: For Research Use Only.