Learn More
Biomatik Corporation Pig Vascular Endothelial cell Growth Factor, VEGF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40841
Encompass_Preferred
Pig Vascular Endothelial Cell Growth Factor, Vegf Elisa Kit for detecting Vascular Endothelial Growth Factor A in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 4.69-300pg/ml. Sensitivity: 1.17pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49151; VEGFA. Target Synonyms: VEGFA; VEGFVascular endothelial growth factor A; VEGF-A; Vascular permeability factor; VPF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9344