Learn More
Biomatik Corporation Human hypoxia-inducible factor 1α (HIF-1α) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40850
Encompass_Preferred
The Human hypoxia-inducible factor 1α (HIF-1α) ELISA Kit allows for quantitative determination of hypoxia-inducible factor 1α (HIF-1α) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q16665. Restriction: For Research Use Only.
Catalog No. 50-149-9346