Learn More
Biomatik Corporation Human Annexin Ⅰ (ANX-Ⅰ) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40857
Encompass_Preferred
The Human Annexin Ⅰ (ANX-Ⅰ) ELISA Kit allows for quantitative determination of annexin A1 in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04083. Restriction: For Research Use Only.
Catalog No. 50-149-9348