Learn More
Biomatik Corporation Rat fibroblast growth factor 23 (FGF23) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40861
Encompass_Preferred
Rat Fibroblast Growth Factor 23 (Fgf23) Elisa Kit for detecting Fibroblast Growth Factor 23 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8VI82; Fgf23. Target Synonyms: Fgf23Fibroblast growth factor 23; FGF-23. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9350