Learn More
Biomatik Corporation Human Interleukin 17A (IL-17A/IL-17) ELISA Kit, 96 tests
Catalog No. 501499360
Encompass_Preferred
The Human Interleukin 17A (IL-17A/IL-17) ELISA Kit allows for quantitative determination interleukin 17A in serum, urine, cell culture supernates and tissue homogenates. Species Reactivity: Human. Detection Range: 6.25 pg/mL-400 pg/mL. Sensitivity: 1.56 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q16552. Restriction: For Research Use Only.
Catalog No. 50-149-9360 Supplier Biomatik Corporation Supplier No. EKC40888
The Human Interleukin 17A (IL-17A/IL-17) ELISA Kit allows for quantitative determination interleukin 17A in serum, urine, cell culture supernates and tissue homogenates. Species Reactivity: Human. Detection Range: 6.25 pg/mL-400 pg/mL. Sensitivity: 1.56 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q16552. Restriction: For Research Use Only.