Learn More
Biomatik Corporation Human desmosine (DES) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40895
Encompass_Preferred
The Human desmosine (DES) ELISA Kit allows for quantitative determination of desmosine (DES) in serum, urine, tissue homogenates. Species Reactivity: Human. Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9365