Learn More
Biomatik Corporation Bovine interleukin 4, IL-4 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40897
Encompass_Preferred
The Bovine interleukin 4, IL-4 ELISA Kit allows for quantitative determination interleukin 4 in serum, plasma, tissue homogenates. Species Reactivity: Bovine. Detection Range: 12.5 pg/mL-800 pg/mL. Sensitivity: 3.12 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P30367. Restriction: For Research Use Only.
Catalog No. 50-149-9366