Learn More
Biomatik Corporation Human Cluster of differentiation 147, CD147 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40912
Encompass_Preferred
The Human Cluster of differentiation 147, CD147 ELISA Kit allows for quantitative determination of basigin (Ok blood group) in serum, plasma. Species Reactivity: Human. Detection Range: 12.5 pg/mL-800 pg/mL. Sensitivity: 3.12 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P35613. Restriction: For Research Use Only.
Catalog No. 50-149-9376