Learn More
Biomatik Corporation Rat asymmetrical dimethylarginine (ADMA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40920
Encompass_Preferred
Rat Asymmetrical Dimethylarginine (Adma) Elisa Kit for detecting Asymmetrical Dimethylarginine(Adma) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.5-300ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9380