Learn More
Biomatik Corporation Mouse fibroblast growth factor 10 (FGF10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40922
Encompass_Preferred
Mouse Fibroblast Growth Factor 10 (Fgf10) Elisa Kit for detecting Fibroblast Growth Factor 10 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O35565; Fgf10. Target Synonyms: Fgf10Fibroblast growth factor 10; FGF-10; Keratinocyte growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9381