Learn More
Biomatik Corporation Bovine Interleukin 8, IL-8 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40925
Encompass_Preferred
The Bovine Interleukin 8, IL-8 ELISA Kit allows for quantitative determination interleukin 8 in serum, plasma, tissue homogenates. Species Reactivity: Bovine. Detection Range: 50 pg/mL-2000 pg/mL. Sensitivity: 50 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P79255. Restriction: For Research Use Only.
Catalog No. 50-149-9383