Learn More
Biomatik Corporation Human Mucin 2 (MUC2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40939
Encompass_Preferred
The Human Mucin 2 (MUC2) ELISA Kit allows for quantitative determination of mucin 2, oligomeric mucus/gel-forming in serum, plasma and cell culture supernates. Species Reactivity: Human. Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q02817. Restriction: For Research Use Only.
Catalog No. 50-149-9386