Learn More
Biomatik Corporation Sheep Interleukin-8, IL-8 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40962
Encompass_Preferred
Sheep Interleukin-8, Il-8 Elisa Kit for detecting Interleukin 8 in serum, plasma, tissue homogenates. Species Reactivity: Sheep (Ovis aries). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36925; CXCL8. Target Synonyms: CXCL8; IL8Interleukin-8; IL-8; C-X-C motif chemokine 8; Chemokine; C-X-C motif) ligand 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9398