Learn More
Biomatik Corporation Mouse N-terminal procollagen propeptide (PNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40983
Encompass_Preferred
Mouse N-Terminal Procollagen Iii Propeptide (Piiinp) Elisa Kit for detecting N-Terminal Procollagen III Propeptide(PIIInp) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: N-terminal procollagen 3 propeptide (P3NP); N-terminal procollagen III propeptide (PIIINP). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9401