Learn More
Biomatik Corporation Pig anti-Mullerian hormone (AMH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40998
Encompass_Preferred
Pig Anti-Mullerian Hormone (Amh) Elisa Kit for detecting Anti-Mullerian Hormone in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.25-50ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P79295; AMH. Target Synonyms: AMH; Muellerian-inhibiting factor; Anti-Muellerian hormone; AMH; Muellerian-inhibiting substance; MIS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9407