Learn More
Biomatik Corporation Rat procalcitonin (PCT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41000
Encompass_Preferred
Rat Procalcitonin (Pct) Elisa Kit for detecting Procalcitonin (Pct) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9408