Learn More
Biomatik Corporation Human Chitinase-3-like Protein 1 (YKL-40/CHI3L1) ELISA Kit, 96 tests
Catalog No. 501499418
Encompass_Preferred
The Human Chitinase-3-like Protein 1 (YKL-40/CHI3L1) ELISA Kit allows for quantitative determination of chitinase 3-like 1 (cartilage glycoprotein-39) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 46.875 pg/mL-3000 pg/mL. Sensitivity: 78.065 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P36222. Restriction: For Research Use Only.
Catalog No. 50-149-9418 Supplier Biomatik Corporation Supplier No. EKC41033
The Human Chitinase-3-like Protein 1 (YKL-40/CHI3L1) ELISA Kit allows for quantitative determination of chitinase 3-like 1 (cartilage glycoprotein-39) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 46.875 pg/mL-3000 pg/mL. Sensitivity: 78.065 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P36222. Restriction: For Research Use Only.