Learn More
Biomatik Corporation Human Transforming Growth factor 3, TGF-3 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41057
Encompass_Preferred
Human Transforming Growth Factor Β3, Tgf-Β3 Elisa Kit for detecting Transforming Growth Factor, Beta 3 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10600; TGFB3. Target Synonyms: ARVD; ARVD1; FLJ16571; LDS5; MGC105479; MGC118722; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; TGF beta3; TGF-beta-3; TGFB 3; Tgfb3; TGFB3_HUMAN; transforming growth factor beta 3; Transforming growth factor beta-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9427