Learn More
Biomatik Corporation Sheep Tumor Necrosis Factor , TNF- ELISA Kit, 96 tests
Catalog No. 501499431
Encompass_Preferred
Sheep Tumor Necrosis Factor Α, Tnf-Α Elisa Kit for detecting Tumor Necrosis Factor (Tnf Superfamily, Member 2) in serum, plasma, tissue homogenates. Species Reactivity: Sheep (Ovis aries). Detection Range: 3.125-50pg/ml. Sensitivity: 3.125pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P23383; TNF. Target Synonyms: TNF; TNFA; TNFSF2; Tumor necrosis factor; Cachectin; TNF-alpha; Tumor necrosis factor ligand superfamily member 2; TNF-a) [Cleaved into: Tumor necrosis factor; membrane form; N-terminal fragment; NTF); Intracellular domain 1; ICD1); Intracellular domain 2; ICD2); C-domain 1; C-domain 2; Tumor necrosis factor; soluble form]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9431 Supplier Biomatik Corporation Supplier No. EKC41070
Sheep Tumor Necrosis Factor Α, Tnf-Α Elisa Kit for detecting Tumor Necrosis Factor (Tnf Superfamily, Member 2) in serum, plasma, tissue homogenates. Species Reactivity: Sheep (Ovis aries). Detection Range: 3.125-50pg/ml. Sensitivity: 3.125pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P23383; TNF. Target Synonyms: TNF; TNFA; TNFSF2; Tumor necrosis factor; Cachectin; TNF-alpha; Tumor necrosis factor ligand superfamily member 2; TNF-a) [Cleaved into: Tumor necrosis factor; membrane form; N-terminal fragment; NTF); Intracellular domain 1; ICD1); Intracellular domain 2; ICD2); C-domain 1; C-domain 2; Tumor necrosis factor; soluble form]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.